Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-2-(prop-1-en-2-yl)aniline 95+% stabilised

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A1501933-100MG A1501933-1G A1501933-250MG
76666-970EA 21 USD InStock
76666-970 76666-972 76666-974
4-Fluoro-2-(prop-1-en-2-yl)aniline 95+% stabilised
4-Fluoro-2-(prop-1-en-2-yl)aniline
Precaución: for Research Use Only


Formula: C₉H₁₀FN
MDL Number: MFCD20699433
CAS Number: 5273-41-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty