Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(OC-6-21)-Tris[3,5-difluoro-2-(2-pyridinyl-κN)phenyl-κC]iridium 97%

Supplier: AMBEED, INC
Warning

A1501940-100MG A1501940-250MG A1501940-25MG A1501940-50MG
76666-976EA 186 USD InStock
76666-976 76666-978 76666-980 76666-982
(OC-6-21)-Tris[3,5-difluoro-2-(2-pyridinyl-κN)phenyl-κC]iridium 97%
(OC-6-21)-Tris[3,5-difluoro-2-(2-pyridinyl-κN)phenyl-κC]iridium
Precaución: for Research Use Only


Formula: C₃₃H₁₈F₆IrN₃
MW: 762.7235 g/mol
CAS Number: 562824-22-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty