Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethyl)benzenesulphonyl chloride 95%

Supplier: AMBEED, INC
Danger

A1502744-25G A1502744-5G
76667-006EA 68 USD InStock
76667-006 77549-874
2-(Trifluoromethyl)benzenesulphonyl chloride 95%
2-(Trifluoromethyl)benzenesulphonyl chloride
Precaución: for Research Use Only


Formula: C₇H₄ClF₃O₂S
MW: 244.62 g/mol
MDL Number: MFCD00051696
CAS Number: 776-04-5
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty