Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,7-Di-tert-butyl-3-(4-methoxyphenyl)benzo[d]oxazol-3-ium tetrafluoroborate 97%

Supplier: AMBEED, INC
Danger

A1503650-1G A1503650-100MG
76667-052EA 267 USD InStock
76667-052 77624-724
5,7-Di-tert-butyl-3-(4-methoxyphenyl)benzo[d]oxazol-3-ium tetrafluoroborate 97%
5,7-Di-tert-butyl-3-(4-methoxyphenyl)benzo[d]oxazol-3-ium tetrafluoroborate
Precaución: for Research Use Only


Formula: C₂₂H₂₈BF₄NO₂
CAS Number: 2750161-86-3
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty