Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethyl-5-methyl-1H-imidazole 95%

Supplier: AMBEED, INC
Warning

A1504958-100G A1504958-25G
76667-124EA 19 USD InStock
76667-124 76667-126
2-Ethyl-5-methyl-1H-imidazole 95%
2-Ethyl-5-methyl-1H-imidazole
Precaución: for Research Use Only


Formula: C₆H₁₀N₂
MDL Number: MFCD23697457
CAS Number: 931-36-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty