Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2,6-Dioxopiperidin-3-yl)-5-iodoisoindoline-1,3-dione 95%

Supplier: AMBEED, INC
Warning

A1505737-1G A1505737-250MG A1505737-100MG A1505737-5G
76667-152EA 391 USD InStock
76667-152 76667-154 77624-746 77624-748
2-(2,6-Dioxopiperidin-3-yl)-5-iodoisoindoline-1,3-dione 95%
2-(2,6-Dioxopiperidin-3-yl)-5-iodoisoindoline-1,3-dione
Precaución: for Research Use Only


Formula: C₁₃H₉IN₂O₄
MDL Number: MFCD31735039
CAS Number: 2509137-01-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty