Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(10,15,20-Tris(4-chlorophenyl)porphyrin-5-yl)phenol 95%

Supplier: AMBEED, INC
Warning

A1508183-100MG A1508183-1G A1508183-250MG A1508183-50MG
76667-316EA 101 USD InStock
76667-316 76667-318 76667-320 76667-322
4-(10,15,20-Tris(4-chlorophenyl)porphyrin-5-yl)phenol 95%
4-(10,15,20-Tris(4-chlorophenyl)porphyrin-5-yl)phenol
Precaución: for Research Use Only


Formula: C₄₄H₂₇Cl₃N₄O
CAS Number: 101395-23-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty