Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(11bS)-N-(2,6-Bis(4-(tert-butyl)phenyl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 95%

Supplier: AMBEED, INC
Warning

A1512895-100MG A1512895-1G A1512895-250MG
76667-424EA 185 USD InStock
76667-424 76667-426 76667-428
(11bS)-N-(2,6-Bis(4-(tert-butyl)phenyl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 95%
(11bS)-N-(2,6-Bis(4-(tert-butyl)phenyl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide
Precaución: for Research Use Only


Formula: C₄₁H₃₇F₃NO₅PS
CAS Number: 2417213-21-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty