Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

15,35,55,75-Tetra-tert-butyl-32,72-dimethoxy-1,3,5,7(1,3)-tetrabenzenacyclooctaphane-12,52-diol 98%

Supplier: AMBEED, INC
Warning

A1519701-1G A1519701-250MG
76667-644EA 100 USD InStock
76667-644 76667-646
15,35,55,75-Tetra-tert-butyl-32,72-dimethoxy-1,3,5,7(1,3)-tetrabenzenacyclooctaphane-12,52-diol 98%
15,35,55,75-Tetra-tert-butyl-32,72-dimethoxy-1,3,5,7(1,3)-tetrabenzenacyclooctaphane-12,52-diol
Precaución: for Research Use Only


Formula: C₄₆H₆₀O₄
MDL Number: MFCD00142526
CAS Number: 122406-45-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty