Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4S,5S)-2-(Benzo[b]thiophen-2-yl)-4,5-diphenyl-4,5-dihydrooxazole 97%

Supplier: AMBEED, INC
Warning

A1519934-100MG A1519934-1G A1519934-250MG
76667-798EA 94 USD InStock
76667-798 77625-182 77625-184
(4S,5S)-2-(Benzo[b]thiophen-2-yl)-4,5-diphenyl-4,5-dihydrooxazole 97%
(4S,5S)-2-(Benzo[b]thiophen-2-yl)-4,5-diphenyl-4,5-dihydrooxazole
Precaución: for Research Use Only


Formula: C₂₃H₁₇NOS
CAS Number: 2828438-83-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty