Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-((2-(Diethylamino)ethyl)carbamoyl)phenyl)boronic acid MIDA ester 98%

Supplier: AMBEED, INC
Danger

A1532954-100MG A1532954-1G A1532954-250MG
76667-976EA 37 USD InStock
76667-976 76667-978 76667-980
(3-((2-(Diethylamino)ethyl)carbamoyl)phenyl)boronic acid MIDA ester 98%
(3-((2-(Diethylamino)ethyl)carbamoyl)phenyl)boronic acid MIDA ester
Precaución: for Research Use Only


Formula: C₁₈H₂₆BN₃O₅
CAS Number: 2828439-48-9
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty