Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-((4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)phenyl)sulfonyl)piperidine 95%

Supplier: AMBEED, INC
Warning

A1535263-100MG A1535263-1G A1535263-250MG
76668-078EA 19 USD InStock
76668-078 76668-080 76668-082
1-((4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)phenyl)sulfonyl)piperidine 95%
1-((4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)phenyl)sulfonyl)piperidine
Precaución: for Research Use Only


Formula: C₁₈H₂₅BF₃NO₄S
MDL Number: MFCD29109976
CAS Number: 2828439-63-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty