Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-6-(trifluoromethyl)benzoate 95+%

Supplier: AMBEED, INC
Warning

A1536130-100MG A1536130-1G A1536130-250MG
76668-092EA 37 USD InStock
76668-092 76668-094 76668-096
Methyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-6-(trifluoromethyl)benzoate 95+%
Methyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-6-(trifluoromethyl)benzoate
Precaución: for Research Use Only


Formula: C₁₅H₁₈BF₃O₄
MW: 330.1072 g/mol
MDL Number: MFCD18756933
CAS Number: 1146214-79-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty