Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Methoxy-5-nitropyridin-3-yl)boronic acid 95%

Supplier: AMBEED, INC
Warning

A1536351-100MG A1536351-1G A1536351-250MG
76668-112EA 19 USD InStock
76668-112 76668-114 76668-116
(2-Methoxy-5-nitropyridin-3-yl)boronic acid 95%
(2-Methoxy-5-nitropyridin-3-yl)boronic acid
Precaución: for Research Use Only


Formula: C₆H₇BN₂O₅
MW: 197.94 g/mol
CAS Number: 2828439-68-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty