Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethylphosphine oxide 90%

Supplier: AMBEED, INC
Danger

A1537079-25G A1537079-1G
76668-124EA 66 USD InStock
76668-124 77592-884
Dimethylphosphine oxide 90%
Dimethylphosphine oxide
Precaución: for Research Use Only


Formula: C₂H₇OP
MDL Number: MFCD02824571
CAS Number: 7211-39-4
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty