Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-((2-Chlorobenzyl)oxy)-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: AMBEED, INC
Warning

A1544833-100MG A1544833-1G A1544833-250MG
76668-502EA 19 USD InStock
76668-502 76668-504 76668-506
2-(4-((2-Chlorobenzyl)oxy)-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(4-((2-Chlorobenzyl)oxy)-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₂₁H₂₆BClO₃
CAS Number: 2828443-98-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty