Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1-Oxo-1,2-dihydroisoquinolin-6-yl)boronic acid 98%

Supplier: AMBEED, INC
Warning

A1564843-1G A1564843-250MG A1564843-5G
76668-560EA 218 USD InStock
76668-560 76668-562 76668-564
(1-Oxo-1,2-dihydroisoquinolin-6-yl)boronic acid 98%
(1-Oxo-1,2-dihydroisoquinolin-6-yl)boronic acid
Precaución: for Research Use Only


Formula: C₉H₈BNO₃
MDL Number: MFCD13189945
CAS Number: 376584-82-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty