Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 97+%

Supplier: AMBEED, INC
Warning

A1617774-100MG A1617774-1G A1617774-250MG
76668-728EA 39 USD InStock
76668-728 76668-730 76668-732
Ethyl 4-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 97+%
Ethyl 4-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Precaución: for Research Use Only


Formula: C₁₅H₂₀BClO₄
MDL Number: MFCD28148050
CAS Number: 1523239-42-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty