Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Bromo-4-fluorophenyl)-N-methylmethanamine 95%

Supplier: AMBEED, INC
Danger

A1618338-100MG A1618338-1G A1618338-250MG
76668-790EA 37 USD InStock
76668-790 76668-792 76668-794
1-(2-Bromo-4-fluorophenyl)-N-methylmethanamine 95%
1-(2-Bromo-4-fluorophenyl)-N-methylmethanamine
Precaución: for Research Use Only


Formula: C₈H₉BrFN
MDL Number: MFCD09810985
CAS Number: 887139-04-0
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty