Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4,5,5-Tetramethyl-2-(4-methyl-2-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane 95%

Supplier: AMBEED, INC
Warning

A1618428-1G A1618428-250MG A1618428-5G
76668-802EA 90 USD InStock
76668-802 76668-804 76668-806
4,4,5,5-Tetramethyl-2-(4-methyl-2-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane 95%
4,4,5,5-Tetramethyl-2-(4-methyl-2-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₁₄H₁₈BF₃O₂
MDL Number: MFCD18729924
CAS Number: 2828445-67-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty