Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

((4-Bromo-3-fluorobenzyl)oxy)(tert-butyl)dimethylsilane 98%

Supplier: AMBEED, INC
Warning

A1627115-1G A1627115-250MG A1627115-5G
76668-902EA 66 USD InStock
76668-902 76668-904 76668-906
((4-Bromo-3-fluorobenzyl)oxy)(tert-butyl)dimethylsilane 98%
((4-Bromo-3-fluorobenzyl)oxy)(tert-butyl)dimethylsilane
Precaución: for Research Use Only


Formula: C₁₃H₂₀BrFOSi
MW: 319.29 g/mol
MDL Number: MFCD31802578
CAS Number: 1017598-60-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty