Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lithium triisopropoxy(4-(trifluoromethyl)pyridin-2-yl)borate 97%

Supplier: AMBEED, INC
Warning

A1639220-1G A1639220-250MG A1639220-5G
76669-132EA 216 USD InStock
76669-132 76669-134 76669-136
Lithium triisopropoxy(4-(trifluoromethyl)pyridin-2-yl)borate 97%
Lithium triisopropoxy(4-(trifluoromethyl)pyridin-2-yl)borate
Precaución: for Research Use Only


Formula: C₁₅H₂₄BF₃LiNO₃
CAS Number: 2095458-45-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty