Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Chloro-3-iodoquinoline 95%

Supplier: AMBEED, INC
Warning

A1669286-100MG A1669286-1G A1669286-250MG
76669-274EA 37 USD InStock
76669-274 76669-276 76669-278
7-Chloro-3-iodoquinoline 95%
7-Chloro-3-iodoquinoline
Precaución: for Research Use Only


Formula: C₉H₅ClIN
MW: 289.50 g/mol
MDL Number: MFCD28667978
CAS Number: 1424245-91-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty