Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(6-(4-Acetylpiperazin-1-yl)pyridin-3-yl)boronic acid hydrochloride 95%

Supplier: AMBEED, INC
Warning

A1672330-1G A1672330-250MG A1672330-5G
76669-390EA 197 USD InStock
76669-390 76669-392 76669-394
(6-(4-Acetylpiperazin-1-yl)pyridin-3-yl)boronic acid hydrochloride 95%
(6-(4-Acetylpiperazin-1-yl)pyridin-3-yl)boronic acid hydrochloride
Precaución: for Research Use Only


Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty