Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Bromo-5-(hydrazinecarbonyl)phenyl)boronic acid hydrochloride 95+%

Supplier: AMBEED, INC
Danger

A1673005-100MG A1673005-1G A1673005-250MG A1673005-5G
76669-450EA 43 USD InStock
76669-450 76669-452 76669-454 76669-456
(3-Bromo-5-(hydrazinecarbonyl)phenyl)boronic acid hydrochloride 95+%
(3-Bromo-5-(hydrazinecarbonyl)phenyl)boronic acid hydrochloride
Precaución: for Research Use Only


Formula: C₇H₉BBrClN₂O₃
UN: 2926
ADR: 4.1,6.1,Ⅱ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty