Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3,4-Bis(benzyloxy)phenyl)boronic acid 98%

Supplier: AMBEED, INC
Warning

A1680751-100MG A1680751-1G A1680751-250MG
76669-590EA 46 USD InStock
76669-590 76669-592 76669-594
(3,4-Bis(benzyloxy)phenyl)boronic acid 98%
(3,4-Bis(benzyloxy)phenyl)boronic acid
Precaución: for Research Use Only


Formula: C₂₀H₁₉BO₄
MDL Number: MFCD33401716
CAS Number: 906665-98-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty