Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-N-(3-chlorophenyl)benzenesulfonamide 98%

Supplier: AMBEED, INC
Warning

A1684694-1G A1684694-250MG A1684694-5G
76669-724EA 66 USD InStock
76669-724 76669-726 76669-728
4-Bromo-N-(3-chlorophenyl)benzenesulfonamide 98%
4-Bromo-N-(3-chlorophenyl)benzenesulphonamide
Precaución: for Research Use Only


Formula: C₁₂H₉BrClNO₂S
MDL Number: MFCD00591726
CAS Number: 7454-56-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty