Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dimethylphenylacetic acid 98%

Supplier: AMBEED, INC
Warning

A629579-10G A629579-1G A629579-25G A629579-5G
76669-800EA 54 USD InStock
76669-800 76669-802 76669-804 76669-806
2,4-Dimethylphenylacetic acid 98%
2,4-Dimethylphenylacetic acid
Precaución: for Research Use Only


Formula: (C₆H₃(CH₃)₂)CH₂COOH
MW: 164.2
MDL Number: MFCD02664685
CAS Number: 6331-04-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty