Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4-Aminophenyl)-1,1,3-trimethyl-2,3-dihydro-1H-inden-5-amine 97%

Supplier: AMBEED, INC
Warning

A1002517-1G A1002517-25G A1002517-5G
76670-752EA 18 USD InStock
76670-752 76670-754 77619-482
3-(4-Aminophenyl)-1,1,3-trimethyl-2,3-dihydro-1H-inden-5-amine 97%
3-(4-Aminophenyl)-1,1,3-trimethyl-2,3-dihydro-1H-inden-5-amine
Precaución: For research use only.


Formula: C₁₈H₂₂N₂
MW: 266.3807 g/mol
MDL Number: MFCD00424120
CAS Number: 54628-90-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty