Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzamidoxime 97%

Supplier: AMBEED, INC
Danger

A1003275-10G A1003275-25G A1003275-5G A1003275-1G
76670-812EA 23 USD InStock
76670-812 76670-814 76670-816 76670-818 77619-498
Benzamidoxime 97%
Benzamidoxime
Precaución: for Research Use Only


Formula: C₇H₈N₂O
MDL Number: MFCD00031485
CAS Number: 613-92-3
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty