Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-BI-DIME 97%

Supplier: AMBEED, INC
Warning

A1005941-100MG A1005941-1G A1005941-250MG A1005941-5G
76670-930EA 63 USD InStock
76670-930 76670-932 76670-934 77619-590
(R)-BI-DIME 97%
(R)-BI-DIME
Precaución: for Research Use Only


Formula: C₁₉H₂₃O₃P
MDL Number: MFCD31630704
CAS Number: 1338454-03-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty