Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Bis((S)-4-methyl-4,5-dihydrooxazol-2-yl)pyridine 97%

Supplier: AMBEED, INC
Warning

A1005971-100MG A1005971-1G A1005971-5G
76670-956EA 11 USD InStock
76670-956 76670-958 77619-596
2,6-Bis((S)-4-methyl-4,5-dihydrooxazol-2-yl)pyridine 97%
2,6-Bis((S)-4-methyl-4,5-dihydrooxazol-2-yl)pyridine
Precaución: for Research Use Only


Formula: C₁₃H₁₅N₃O₂
MDL Number: MFCD31706427
CAS Number: 189014-95-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty