Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methyl-4-(propan-2-yl)aniline hydrochloride 95%

Supplier: AMBEED, INC
Warning

A1014836-100MG A1014836-1G A1014836-250MG A1014836-25G A1014836-5G
76671-092EA 9 USD InStock
76671-092 76671-094 76671-096 76671-098 76671-100
N-Methyl-4-(propan-2-yl)aniline hydrochloride 95%
N-Methyl-4-(propan-2-yl)aniline hydrochloride
Precaución: for Research Use Only


Formula: C₁₀H₁₆ClN
MW: 185.6937 g/mol
MDL Number: MFCD22378668
CAS Number: 1394042-73-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty