Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Diethyl-2-phenylbutanamide 95%

Supplier: AMBEED, INC
Warning

A1018627-1G A1018627-250MG A1018627-5G
76671-108EA 84 USD InStock
76671-108 76671-110 76671-112
N,N-Diethyl-2-phenylbutanamide 95%
N,N-Diethyl-2-phenylbutanamide
Precaución: for Research Use Only


Formula: C₁₄H₂₁NO
MDL Number: MFCD01356286
CAS Number: 92321-53-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty