Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 1-(prop-2-enoyl)piperidine-4-carboxylate 95%

Supplier: AMBEED, INC
Warning

A1022537-100MG A1022537-1G A1022537-250MG A1022537-50MG
76671-120EA 26 USD InStock
76671-120 76671-122 76671-124 76671-126
Sodium 1-(prop-2-enoyl)piperidine-4-carboxylate 95%
Sodium 1-(prop-2-enoyl)piperidine-4-carboxylate
Precaución: for Research Use Only


Formula: C₉H₁₂NNaO₃
MDL Number: MFCD22421872
CAS Number: 1423031-89-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty