Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Phenyl-1H-indole 95%

Supplier: AMBEED, INC
Warning

A1025341-100MG A1025341-1G A1025341-250MG
76671-142EA 49 USD InStock
76671-142 76671-144 76671-146
4-Phenyl-1H-indole 95%
4-Phenyl-1H-indole
Precaución: for Research Use Only


Formula: C₁₄H₁₁N
MW: 193.2400 g/mol
MDL Number: MFCD13177532
CAS Number: 35577-92-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty