Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Ethyl-N-isopropylnitrous amide 95%

Supplier: AMBEED, INC
Danger

A1036012-100MG A1036012-250MG
76671-290EA 34 USD InStock
76671-290 77619-704
N-Ethyl-N-isopropylnitrous amide 95%
N-Ethyl-N-isopropylnitrous amide
Precaución: for Research Use Only


Formula: C₅H₁₂N₂O
MW: 116.1616 g/mol
MDL Number: MFCD01730482
CAS Number: 16339-04-1
UN: 2810
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty