Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-hydroxy-3,3-dimethylbutanoate 95%

Supplier: AMBEED, INC
Danger

A1037062-1G A1037062-100MG
76671-296EA 107 USD InStock
76671-296 77619-706
Methyl 2-hydroxy-3,3-dimethylbutanoate 95%
Methyl 2-hydroxy-3,3-dimethylbutanoate
Precaución: for Research Use Only


Formula: C₇H₁₄O₃
MDL Number: MFCD16693888
CAS Number: 121129-31-5
UN: 1987
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty