Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dimethoxybenzene-1-sulfonyl chloride 95%

Supplier: AMBEED, INC
Danger

A1040395-1G A1040395-5G
76671-360EA 42 USD InStock
76671-360 76671-362
2,6-Dimethoxybenzene-1-sulfonyl chloride 95%
2,6-Dimethoxybenzene-1-sulfonyl chloride
Precaución: for Research Use Only


Formula: C₈H₉ClO₄S
MW: 236.6727 g/mol
MDL Number: MFCD18395734
CAS Number: 145980-89-8
UN: 3261
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty