Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyano-2-ethylbutanoic acid 95%

Supplier: AMBEED, INC
Warning

A1040948-1G A1040948-250MG A1040948-5G
76671-372EA 212 USD InStock
76671-372 76671-374 76671-376
2-Cyano-2-ethylbutanoic acid 95%
2-Cyano-2-ethylbutanoic acid
Precaución: for Research Use Only


Formula: C₇H₁₁NO₂
MDL Number: MFCD00152061
CAS Number: 4386-07-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty