Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl N-methyl-N-[4-(methylamino)butyl]carbamate 95%

Supplier: AMBEED, INC
Danger

A1055281-100MG A1055281-1G
76671-518EA 154 USD InStock
76671-518 77619-752
tert-Butyl N-methyl-N-[4-(methylamino)butyl]carbamate 95%
tert-Butyl N-methyl-N-[4-(methylamino)butyl]carbamate
Precaución: for Research Use Only


Formula: C₁₁H₂₄N₂O₂
MW: 216.3205 g/mol
MDL Number: MFCD24472025
CAS Number: 122632-07-9
UN: 1760
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty