Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-[4-Fluoro-3-(prop-2-enamido)phenyl]-1-(2-fluorophenyl)-1H-pyrazole-4-carboxamide 98%

Supplier: AMBEED, INC
Danger

A1060682-10MG A1060682-1MG A1060682-50MG
76671-614EA 113 USD InStock
76671-614 76671-616 76671-618
N-[4-Fluoro-3-(prop-2-enamido)phenyl]-1-(2-fluorophenyl)-1H-pyrazole-4-carboxamide 98%
N-[4-Fluoro-3-(prop-2-enamido)phenyl]-1-(2-fluorophenyl)-1H-pyrazole-4-carboxamide
Precaución: for Research Use Only


Formula: C₁₉H₁₄F₂N₄O₂
MDL Number: MFCD31665468
CAS Number: 1808714-73-9
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty