Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S)-2-Amino-5,5,5-trifluoropentanoic acid hydrochloride 95%

Supplier: AMBEED, INC
Warning

A1060748-100MG A1060748-250MG
76671-620EA 58 USD InStock
76671-620 76671-622
(2S)-2-Amino-5,5,5-trifluoropentanoic acid hydrochloride 95%
(2S)-2-Amino-5,5,5-trifluoropentanoic acid hydrochloride
Precaución: for Research Use Only


Formula: C₅H₉ClF₃NO₂
MDL Number: MFCD31666195
CAS Number: 1680174-93-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty