Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(2-(1H-Indol-3-yl)ethyl)-2-aminoacetamide 95%

Supplier: AMBEED, INC
Warning

A1061187-100MG A1061187-1G A1061187-250MG
76671-626EA 65 USD InStock
76671-626 76671-628 76671-630
N-(2-(1H-Indol-3-yl)ethyl)-2-aminoacetamide 95%
N-(2-(1H-Indol-3-yl)ethyl)-2-aminoacetamide
Precaución: for Research Use Only


Formula: C₁₂H₁₅N₃O
MDL Number: MFCD08282638
CAS Number: 122902-82-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty