Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(Pyridin-2-yl)prop-2-enamide 95%

Supplier: AMBEED, INC
Warning

A1061900-100MG A1061900-1G A1061900-250MG
76671-640EA 28 USD InStock
76671-640 76671-642 76671-644
N-(Pyridin-2-yl)prop-2-enamide 95%
N-(Pyridin-2-yl)prop-2-enamide
Precaución: for Research Use Only


Formula: C₈H₈N₂O
MDL Number: MFCD00596317
CAS Number: 40000-69-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty