Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[4-(Ethenyloxy)phenyl]methanol 95% stabilized

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A1062237-100MG A1062237-1G A1062237-250MG
76671-662EA 88 USD InStock
76671-662 76671-664 76671-666
[4-(Ethenyloxy)phenyl]methanol 95% stabilized
[4-(Ethenyloxy)phenyl]methanol
Precaución: for Research Use Only


Formula: C₉H₁₀O₂
MDL Number: MFCD28064182
CAS Number: 77089-07-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty