Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5H,6H,7H,8H-Imidazo[1,5-a]pyridine-6-carboxylic acid hydrochloride 95%

Supplier: AMBEED, INC
Warning

A1067279-100MG A1067279-250MG A1067279-50MG
76671-776EA 227 USD InStock
76671-776 76671-778 76671-780
5H,6H,7H,8H-Imidazo[1,5-a]pyridine-6-carboxylic acid hydrochloride 95%
5H,6H,7H,8H-Imidazo[1,5-a]pyridine-6-carboxylic acid hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₁ClN₂O₂
MDL Number: MFCD28246531
CAS Number: 139183-92-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty