Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrazine-2,5-dicarbonitrile 98%

Supplier: AMBEED, INC
Danger

A1067383-1G A1067383-5G A1067383-250MG
76671-782EA 343 USD InStock
76671-782 76671-784 77619-774
Pyrazine-2,5-dicarbonitrile 98%
Pyrazine-2,5-dicarbonitrile
Precaución: for Research Use Only


Formula: C₆H₂N₄
MDL Number: MFCD15144323
CAS Number: 31722-48-2
UN: 3439
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty