Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Chlorosulfonyl)-2-methylbenzoyl chloride 95%

Supplier: AMBEED, INC
Danger

A1068692-100MG A1068692-1G A1068692-250MG
76671-818EA 122 USD InStock
76671-818 76671-820 76671-822
5-(Chlorosulfonyl)-2-methylbenzoyl chloride 95%
5-(Chlorosulfonyl)-2-methylbenzoyl chloride
Precaución: for Research Use Only


Formula: C₈H₆Cl₂O₃S
MDL Number: MFCD28397465
CAS Number: 1426437-45-7
UN: 2927
ADR: 6.1,8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty