Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitro-2,1,3-benzoselenadiazole 95%

Supplier: AMBEED, INC
Danger

A1070530-100MG A1070530-1G A1070530-250MG
76671-854EA 48 USD InStock
76671-854 76671-856 76671-858
4-Nitro-2,1,3-benzoselenadiazole 95%
4-Nitro-2,1,3-benzoselenadiazole
Precaución: for Research Use Only


Formula: C₆H₃N₃O₂Se
MDL Number: MFCD00182136
CAS Number: 20718-41-6
UN: 3283
ADR: 6.1,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty